Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001122963.1 | NP_001116435.1 | 5 | 1662 | |
NM_028487.4 | NP_082763.3 | 5 | 1662 | ||
GenBank mRNA | AK082704.1 | - | 5 | 1480 | |
AK135728.1 | - | 5 | 1593 | ||
AK136894.1 | - | 5 | 1441 | ||
AK145318.1 | - | 5 | 1651 | ||
AK151283.1 | - | 4 | 883 | ||
AK161928.1 | - | 5 | 1450 | ||
AK163370.1 | - | 5 | 1510 | ||
AK163390.1 | - | 5 | 1664 | ||
AK169406.1 | - | 5 | 1457 | ||
AY382529.1 | - | 5 | 1460 | ||
BC067075.1 | - | 5 | 1436 | 30614988 | |
BC076635.1 | - | 5 | 1492 | ||
BC094888.1 | - | 5 | 1455 | 30628044 | |
BC096442.1 | - | 5 | 1479 |