Gene Transcripts
- Gene Symbol
- Lima1
- Entrez Gene ID
- 65970
- Gene Name
- LIM domain and actin binding 1
- Gene Aliases
- Eplin
- Chromosome Location
- Chr. 15: 99778468 - 99875456 on Build GRCm38
- UniGene ID
- Mm.33207
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001113545.1 |
NP_001107017.1
|
10, 11 |
1424 |
|
NM_023063.4 |
NP_075550.2
|
7, 8 |
1296 |
|
GenBank mRNA |
AF307844.1 |
- |
7, 8 |
1219 |
|
AF307845.1 |
- |
10, 11 |
1378 |
|
AK028186.1 |
- |
7, 8 |
1237 |
|
AK031698.1 |
- |
7, 8 |
1240 |
|
AK049350.1 |
- |
10, 11 |
1374 |
|
AK085065.1 |
- |
7, 8 |
1290 |
|
AK132314.1 |
- |
7, 8 |
1242 |
|
AK167220.1 |
- |
7, 8 |
856 |
|
BC031490.1 |
- |
10, 11 |
1350 |
3498138 |
View Full Product Details