Gene Transcripts

Gene Symbol
Stag1
Entrez Gene ID
20842
Gene Name
stromal antigen 1
Gene Aliases
AU045003, SA-1, Scc3
Chromosome Location
Chr. 9: 100597709 - 100958544 on Build GRCm38
UniGene ID
Mm.42135
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009282.3 NP_033308.2 25 2829
GenBank mRNA BC062954.1 - 25 2829
BC069841.1 - 26 2757
Z75332.1 - 25 2878

View Full Product Details