Gene Transcripts

Gene Symbol
Lmo7
Entrez Gene ID
380928
Gene Name
LIM domain only 7
Gene Aliases
C78582, FBXO20, Gm914, mKIAA0858
Chromosome Location
Chr. 14: 101729917 - 101934708 on Build GRCm38
UniGene ID
Mm.218981
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_201529.2 NP_963287.2 28 5083
GenBank mRNA AK047836.1 - 4 2012
AK129231.1 - 25 4868
AK137694.1 - 19 3428
BC082553.1 - 26 5098
BC145011.1 - 21 3567 9053559
EF071857.1 - 28 5054

View Full Product Details