Gene Transcripts

Gene Symbol
Parp8
Entrez Gene ID
52552
Gene Name
poly (ADP-ribose) polymerase family, member 8
Gene Aliases
2810430O08Rik, ARTD16, D13Ertd275e
Chromosome Location
Chr. 13: 116850842 - 117025913 on Build GRCm38
UniGene ID
Mm.274466
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081009.1 NP_001074478.1 7 555
GenBank mRNA AK149889.1 - 6 522
AK149918.1 - 6 522
AK150113.1 - 6 468
AK150208.1 - 6 522
AK150732.1 - 6 400
AK151510.1 - 6 400
AK152919.1 - 6 400
AK153169.1 - 6 400
AK153270.1 - 6 400
BC021881.1 - 6 525 3710450

View Full Product Details