Gene Transcripts

Gene Symbol
Trpc6
Entrez Gene ID
22068
Gene Name
transient receptor potential cation channel, subfamily C, member 6
Gene Aliases
AV025995, LLHWJM002, LLHWJM003, LLHWJM004, TRP-6, Trrp6, mtrp6
Chromosome Location
Chr. 9: 8543932 - 8680752 on Build GRCm38
UniGene ID
Mm.325086
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282086.1 NP_001269015.1 2 1111
NM_001282087.1 NP_001269016.1 2 1111
NM_013838.2 NP_038866.2 2 1111
GenBank mRNA AF057748.1 - 2 680
AK133864.1 - 2 1011
AK137447.1 - 1 341
BC067391.1 - 2 1057
BC068310.1 - 2 1057
BC141131.1 - 2 982 9055743
U49069.1 - 2 1111

View Full Product Details