Gene Transcripts

Gene Symbol
Vmn2r36
Entrez Gene ID
100042653
Gene Name
vomeronasal 2, receptor 36
Gene Aliases
100042724, Gm3991
Chromosome Location
Chr. 7: 7876653 - 7902462 on Build GRCm38
UniGene ID
Mm.465202
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105068.1 NP_001098538.1 2 440

Gene Symbol
Vmn2r40
Entrez Gene ID
100042781
Gene Name
vomeronasal 2, receptor 40
Gene Aliases
Chromosome Location
Chr. 7: 8907734 - 8931402 on Build GRCm38
UniGene ID
Mm.469893
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105072.1 NP_001098542.1 2 440

Gene Symbol
Vmn2r41
Entrez Gene ID
100042848
Gene Name
vomeronasal 2, receptor 41
Gene Aliases
Chromosome Location
Chr. 7: 8137905 - 8161551 on Build GRCm38
UniGene ID
Mm.469892
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105073.1 NP_001098543.1 2 440

Gene Symbol
Vmn2r48
Entrez Gene ID
625580
Gene Name
vomeronasal 2, receptor 48
Gene Aliases
EG625580
Chromosome Location
Chr. 7: 9927624 - 9953585 on Build GRCm38
UniGene ID
Mm.469893
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105152.1 NP_001098622.1 2 449

View Full Product Details