Gene Transcripts

Gene Symbol
Cypt4
Entrez Gene ID
235067
Gene Name
cysteine-rich perinuclear theca 4
Gene Aliases
1700009B20Rik, Ckt1r2
Chromosome Location
Chr. 9: 24625185 - 24625828 on Build GRCm38
UniGene ID
Mm.437189
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173412.2 NP_775588.2 1 561
GenBank mRNA AK005791.1 - 1 554
BC060952.1 - 1 550 6772242
BC147783.1 - 1 645 9087502

View Full Product Details