Gene Transcripts

Gene Symbol
Ssxb2
Entrez Gene ID
387132
Gene Name
synovial sarcoma, X member B, breakpoint 2
Gene Aliases
Chromosome Location
Chr. X: 8454343 - 8461726 on Build GRCm38
UniGene ID
Mm.379565
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001450.4 NP_001001450.1 5 570
NM_001134226.1 NP_001127698.1 4 393
GenBank mRNA AK076879.1 - 5 571
AY244793.1 - 5 571
BC048441.1 - 4 393 6773922
BC089478.1 - 4 393 6743842

View Full Product Details