Gene Transcripts

Gene Symbol
Gys2
Entrez Gene ID
232493
Gene Name
glycogen synthase 2
Gene Aliases
BC021322, LGS
Chromosome Location
Chr. 6: 142422612 - 142473124 on Build GRCm38
UniGene ID
Mm.275975
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145572.2 NP_663547.2 16 2363
GenBank mRNA AK138992.1 - 16 2071
AK149391.1 - 16 2088
BC021322.1 - 16 2224 5051685
BC158081.1 - 16 2224 9007373
BX516187.1 - 1 65

View Full Product Details