Gene Transcripts

Gene Symbol
Vmn2r39
Entrez Gene ID
545909
Gene Name
vomeronasal 2, receptor 39
Gene Aliases
EG545909
Chromosome Location
Chr. 7: 9014750 - 9030682 on Build GRCm38
UniGene ID
Mm.469894
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105071.1 NP_001098541.1 4 1370

View Full Product Details