Gene Transcripts

Gene Symbol
Col2a1
Entrez Gene ID
12824
Gene Name
collagen, type II, alpha 1
Gene Aliases
Col2, Col2a, Col2a-1, Del1, Dmm, Lpk, M100856, Rgsc856
Chromosome Location
Chr. 15: 97975602 - 98004724 on Build GRCm38
UniGene ID
Mm.2423
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113515.2 NP_001106987.2 3, 4 371
NM_031163.3 NP_112440.2 4, 5 575
GenBank mRNA AK132311.1 - 3, 4 292
BC051383.1 - 3, 4 282 6312433
BC052326.1 - 3, 4 261 6315379
BC082331.1 - 4, 5 508 30537724

View Full Product Details