Gene Transcripts

Gene Symbol
Phip
Entrez Gene ID
83946
Gene Name
pleckstrin homology domain interacting protein
Gene Aliases
2810004D21Rik, 4632404O06Rik, Ndrp, Wdr11
Chromosome Location
Chr. 9: 82866159 - 82975700 on Build GRCm38
UniGene ID
Mm.221688
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081216.1 NP_001074685.1 21 2585
GenBank mRNA AB049460.1 - 21 2504
AJ303103.1 - 21 2481
AK049736.1 - 21 2521
AK162189.1 - 2 155
BC009146.1 - 12 1459
BC022102.1 - 21 2523
BC037036.1 - 21 2514
BC039765.1 - 21 2537
BC049950.1 - 21 2515
BC068022.1 - 13 1576
BC108423.1 - 598

View Full Product Details