Gene Transcripts

Gene Symbol
Rnaset2a
Entrez Gene ID
100037283
Gene Name
ribonuclease T2A
Gene Aliases
0610007O07Rik, 4833423A10Rik, 4930532K22Rik, C79750, RNASE6PL, Rnaset2
Chromosome Location
Chr. 17: 8128592 - 8147827 on Build GRCm38
UniGene ID
Mm.426736
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083938.2 NP_001077407.1 4 352
GenBank mRNA AK014754.1 - 1 195
AK018722.1 - 4 352
AK019234.1 - 4 344
BC031496.1 - 4 332 3156549
BC089534.1 - 4 661 6703307

Gene Symbol
Rnaset2b
Entrez Gene ID
68195
Gene Name
ribonuclease T2B
Gene Aliases
0610007O07Rik, 4833423A10Rik, 4930532K22Rik, C79750, RNASE6PL, Rnaset2
Chromosome Location
Chr. 17: 6978860 - 6998193 on Build GRCm38
UniGene ID
Mm.181237
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026611.2 NP_080887.1 4 396
GenBank mRNA AK029149.1 - 350
AK084454.1 - 356
BC100330.1 - 4 321 30527517

View Full Product Details