Gene Transcripts
- Gene Symbol
- Hist1h3i
- Entrez Gene ID
- 319153
- Gene Name
- histone cluster 1, H3i
- Gene Aliases
- H3-H
- Chromosome Location
- Chr. 13: 21782915 - 21783397 on Build GRCm38
- UniGene ID
- Mm.390558
- Species
- Mus musculus
- Gene Symbol
- Hist1h3a
- Entrez Gene ID
- 360198
- Gene Name
- histone cluster 1, H3a
- Gene Aliases
- Chromosome Location
- Chr. 13: 23761885 - 23762386 on Build GRCm38
- UniGene ID
- Mm.221301
- Species
- Mus musculus
- Gene Symbol
- Hist1h3b
- Entrez Gene ID
- 319150
- Gene Name
- histone cluster 1, H3b
- Gene Aliases
- H3-53
- Chromosome Location
- Chr. 13: 23752380 - 23752840 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_178203.2 |
NP_835510.1
|
1 |
229 |
|
- Gene Symbol
- Hist1h3c
- Entrez Gene ID
- 319148
- Gene Name
- histone cluster 1, H3c
- Gene Aliases
- H3-143
- Chromosome Location
- Chr. 13: 23745042 - 23745521 on Build GRCm38
- UniGene ID
- Mm.262517
- Species
- Mus musculus
- Gene Symbol
- Hist1h3d
- Entrez Gene ID
- 319149
- Gene Name
- histone cluster 1, H3d
- Gene Aliases
- H3-b
- Chromosome Location
- Chr. 13: 23575763 - 23576266 on Build GRCm38
- UniGene ID
- Mm.347699
- Species
- Mus musculus
- Gene Symbol
- Hist1h3e
- Entrez Gene ID
- 319151
- Gene Name
- histone cluster 1, H3e
- Gene Aliases
- H3-f, H3.1
- Chromosome Location
- Chr. 13: 23561896 - 23562365 on Build GRCm38
- UniGene ID
- Mm.261658
- Species
- Mus musculus
- Gene Symbol
- Hist1h3f
- Entrez Gene ID
- 260423
- Gene Name
- histone cluster 1, H3f
- Gene Aliases
- H3.2-221, Hist1
- Chromosome Location
- Chr. 13: 23544052 - 23544954 on Build GRCm38
- UniGene ID
- Mm.221389
- Species
- Mus musculus
- Gene Symbol
- Hist1h3g
- Entrez Gene ID
- 97908
- Gene Name
- histone cluster 1, H3g
- Gene Aliases
- H3.1-221, M32460
- Chromosome Location
- Chr. 13: 23535418 - 23535909 on Build GRCm38
- UniGene ID
- Mm.261657
- Species
- Mus musculus
- Gene Symbol
- Hist1h3h
- Entrez Gene ID
- 319152
- Gene Name
- histone cluster 1, H3h
- Gene Aliases
- H3-291
- Chromosome Location
- Chr. 13: 21717628 - 21718115 on Build GRCm38
- UniGene ID
- Mm.377874
- Species
- Mus musculus
- Gene Symbol
- Hist2h3b
- Entrez Gene ID
- 319154
- Gene Name
- histone cluster 2, H3b
- Gene Aliases
- H3-615, H3-616
- Chromosome Location
- Chr. 3: 96268654 - 96269155 on Build GRCm38
- UniGene ID
- Mm.383293
- Species
- Mus musculus
- Gene Symbol
- Hist2h3c1
- Entrez Gene ID
- 15077
- Gene Name
- histone cluster 2, H3c1
- Gene Aliases
- BE691662, H3-615, H3f2, Hist2h3ca1
- Chromosome Location
- Chr. 3: 96246685 - 96247348 on Build GRCm38
- UniGene ID
- Mm.422680
- Species
- Mus musculus
- Gene Symbol
- Hist2h3c2
- Entrez Gene ID
- 97114
- Gene Name
- histone cluster 2, H3c2
- Gene Aliases
- H3-614, H3.2-614, Hist2h3c2-ps, Hist2h3ca1, M32461
- Chromosome Location
- Chr. 3: 96238479 - 96239127 on Build GRCm38
- UniGene ID
- Mm.358797
- Species
- Mus musculus
View Full Product Details