Gene Transcripts

Gene Symbol
KLHL18
Entrez Gene ID
23276
Gene Name
kelch like family member 18
Gene Aliases
Chromosome Location
Chr. 3: 47282840 - 47346816 on Build GRCh38
UniGene ID
Hs.517946
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025010.4 NP_079286.2 4 703
XM_005265001.3 XP_005265058.2 4 603
XM_005265002.4 XP_005265059.2 4 603
XM_005265003.3 XP_005265060.2 4 603
XM_006713079.1 XP_006713142.1 4 559
XM_017006033.1 XP_016861522.1 3 466
XM_017006034.1 XP_016861523.1 3 466
XM_017006035.1 XP_016861524.1 3 466
GenBank mRNA AB018338.1 - 2 251
AB062478.1 - 4 875
AK023765.1 - 4 598
AK291477.1 - 4 597
AK295292.1 - 3 465
AY714879.1 - 1 130
BC032620.1 - 4 515 5578173
BX537953.1 - 4 594

View Full Product Details