Gene Transcripts
- Gene Symbol
- Hist1h4j
- Entrez Gene ID
- 319159
- Gene Name
- histone cluster 1, H4j
- Gene Aliases
- Chromosome Location
- Chr. 13: 21735096 - 21735407 on Build GRCm38
- UniGene ID
- Mm.486099
- Species
- Mus musculus
- Gene Symbol
- Hist1h4b
- Entrez Gene ID
- 326620
- Gene Name
- histone cluster 1, H4b
- Gene Aliases
- Chromosome Location
- Chr. 13: 23756937 - 23757386 on Build GRCm38
- UniGene ID
- Mm.261642
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC132186.1 |
- |
1 |
169 |
|
BC132212.1 |
- |
1 |
169 |
40130489 |
- Gene Symbol
- Hist1h4h
- Entrez Gene ID
- 69386
- Gene Name
- histone cluster 1, H4h
- Gene Aliases
- 1700024H08Rik
- Chromosome Location
- Chr. 13: 23531044 - 23531478 on Build GRCm38
- UniGene ID
- Mm.158272
- Species
- Mus musculus
- Gene Symbol
- Hist1h4m
- Entrez Gene ID
- 100041230
- Gene Name
- histone cluster 1, H4m
- Gene Aliases
- Gm11275, OTTMUSG00000000446
- Chromosome Location
- Chr. 13: 21811746 - 21812150 on Build GRCm38
- UniGene ID
- Mm.442307
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC139425.1 |
- |
|
147 |
|
BC139437.1 |
- |
|
147 |
8862459 |
View Full Product Details