Gene Transcripts

Gene Symbol
Mcpt8l2
Entrez Gene ID
408240
Gene Name
mast cell protease 8-like 2
Gene Aliases
Chromosome Location
Chr. : - on Build
UniGene ID
Rn.154796
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135010.1 NP_001128482.1 3 326
GenBank mRNA FQ229939.1 - 3 388

View Full Product Details