Gene Transcripts

Gene Symbol
ANKS1A
Entrez Gene ID
23294
Gene Name
ankyrin repeat and sterile alpha motif domain containing 1A
Gene Aliases
ANKS1
Chromosome Location
Chr. 6: 34889261 - 35097994 on Build GRCh38
UniGene ID
Hs.656492
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015245.2 NP_056060.2 21 3198
XM_005248964.3 XP_005249021.1 22 3249
XM_006715036.2 XP_006715099.1 22 3252
XM_011514431.2 XP_011512733.1 22 3252
XM_011514432.2 XP_011512734.1 22 3249
XM_011514433.1 XP_011512735.1 22 3252
XM_011514434.2 XP_011512736.1 21 3189
XM_011514435.2 XP_011512737.1 21 3186
XM_011514436.2 XP_011512738.1 22 6821
XM_011514437.2 XP_011512739.1 12 3077
XM_017010593.1 XP_016866082.1 12 1675
GenBank mRNA BC031934.1 - 23 2864
BC049213.1 - 10 1041
BC132832.1 - 21 3135 40146854
BQ216321.1 - 6 581
BX094969.1 - 4 327
D86982.1 - 21 3195

View Full Product Details