Gene Transcripts

Gene Symbol
Slpil2
Entrez Gene ID
408229
Gene Name
antileukoproteinase-like 2
Gene Aliases
Chromosome Location
Chr. 3: 160736892 - 160738920 on Build Rnor_6.0
UniGene ID
Rn.198174
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008872.1 NP_001008872.1 3 351
XM_008762532.1 XP_008760754.1 3 351

Gene Symbol
LOC103694893
Entrez Gene ID
103694893
Gene Name
antileukoproteinase-like
Gene Aliases
Chromosome Location
Chr. Un|NW_007906879: 85887 - 87915 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008773874.1 XP_008772096.1 3 351

View Full Product Details