Gene Transcripts

Gene Symbol
March9
Entrez Gene ID
679272
Gene Name
membrane-associated ring finger (C3HC4) 9
Gene Aliases
March-IX, Rnf179
Chromosome Location
Chr. 7: 70341989 - 70349636 on Build Rnor_6.0
UniGene ID
Rn.216720
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100601.1 NP_001094071.1 3 592
XM_008765422.1 XP_008763644.1 4 1567
GenBank mRNA AB048842.1 - 3 592

View Full Product Details