Gene Transcripts

Gene Symbol
Vom2r34
Entrez Gene ID
691767
Gene Name
vomeronasal 2 receptor, 34
Gene Aliases
Chromosome Location
Chr. : - on Build
UniGene ID
Rn.217636
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099657.1 NP_001093127.1 3, 4 1030

View Full Product Details