Gene Transcripts

Gene Symbol
WDR7
Entrez Gene ID
23335
Gene Name
WD repeat domain 7
Gene Aliases
TRAG
Chromosome Location
Chr. 18: 56651343 - 57029805 on Build GRCh38
UniGene ID
Hs.465213
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015285.2 NP_056100.2 12 1599
NM_052834.2 NP_443066.2 12 1599
XM_006722431.2 XP_006722494.1 12 1531
XM_011525888.1 XP_011524190.1 12 1641
XM_017025682.1 XP_016881171.1 12 1504
GenBank mRNA AB011113.2 - 12 1562
AF188125.1 - 11 1388
AK057141.1 - 12 1622
AY099325.1 - 11 1388
BC150282.1 - 12 1562 9007268

View Full Product Details