Gene Transcripts

Gene Symbol
Hcrt
Entrez Gene ID
25723
Gene Name
hypocretin
Gene Aliases
orexin-A
Chromosome Location
Chr. 10: 88669216 - 88670430 on Build Rnor_6.0
UniGene ID
Rn.7628
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013179.2 NP_037311.1 2 503
GenBank mRNA AF019565.1 - 2 508
AF041241.1 - 2 507

View Full Product Details