Gene Transcripts

Gene Symbol
Dmrta2
Entrez Gene ID
313471
Gene Name
DMRT-like family A2
Gene Aliases
Chromosome Location
Chr. 5: 129754295 - 129760045 on Build Rnor_6.0
UniGene ID
Rn.20294
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107951.1 NP_001101421.1 3 2325
XM_006238589.1 XP_006238651.1 3 2324

View Full Product Details