Gene Transcripts

Gene Symbol
Twf2
Entrez Gene ID
684352
Gene Name
twinfilin actin-binding protein 2
Gene Aliases
Chromosome Location
Chr. 8: 114897011 - 114909531 on Build Rnor_6.0
UniGene ID
Rn.216422
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135238.1 NP_001128710.1 8, 9 1027
GenBank mRNA BC158614.1 - 8, 9 975

View Full Product Details