Gene Transcripts

Gene Symbol
Gnrh1
Entrez Gene ID
25194
Gene Name
gonadotropin-releasing hormone 1 (luteinizing-releasing hormone)
Gene Aliases
Gnrh, Gnrha, Lhrh, Rgnrhg1
Chromosome Location
Chr. 15: 44441856 - 44446064 on Build Rnor_6.0
UniGene ID
Rn.159986
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012767.2 NP_036899.1 4 427
XM_006252096.2 XP_006252158.1 3 428
GenBank mRNA M12579.1 - 4 427

View Full Product Details