Gene Transcripts

Gene Symbol
Ceacam6
Entrez Gene ID
100125369
Gene Name
carcinoembryonic antigen-related cell adhesion molecule 6
Gene Aliases
Chromosome Location
Chr. 1: 81683500 - 81701230 on Build Rnor_6.0
UniGene ID
Rn.122781
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB434131.1 - 2 318
BC078962.1 - 2 557 7111028

Gene Symbol
LOC103689938
Entrez Gene ID
103689938
Gene Name
carcinoembryonic antigen-related cell adhesion molecule 1-like
Gene Aliases
Chromosome Location
Chr. 1: 83043478 - 83061421 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008759078.1 XP_008757300.1 600

View Full Product Details