Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017151.2 | NP_058847.1 | 3 | 123 | |
GenBank mRNA | AY390387.1 | - | 2 | 51 | |
FQ211784.1 | - | 3 | 123 | ||
FQ217117.1 | - | 3 | 124 | ||
FQ217914.1 | - | 3 | 123 | ||
FQ220999.1 | - | 3 | 123 | ||
FQ221086.1 | - | 3 | 124 | ||
FQ221189.1 | - | 3 | 123 | ||
FQ221217.1 | - | 3 | 124 | ||
FQ221800.1 | - | 3 | 123 | ||
FQ222134.1 | - | 3 | 123 | ||
FQ222137.1 | - | 3 | 124 | ||
FQ222200.1 | - | 3 | 123 | ||
FQ222233.1 | - | 3 | 122 | ||
FQ222539.1 | - | 3 | 123 | ||
FQ222610.1 | - | 3 | 123 | ||
FQ222928.1 | - | 3 | 123 | ||
FQ223014.1 | - | 2 | 125 | ||
FQ223311.1 | - | 3 | 124 | ||
FQ223478.1 | - | 3 | 126 | ||
FQ223811.1 | - | 3 | 123 | ||
FQ223860.1 | - | 3 | 119 | ||
FQ224530.1 | - | 3 | 123 | ||
FQ228367.1 | - | 3 | 131 | ||
FQ228405.1 | - | 3 | 126 | ||
M19393.1 | - | 3 | 123 |