Gene Transcripts

Gene Symbol
Trappc6a
Entrez Gene ID
680465
Gene Name
trafficking protein particle complex 6A
Gene Aliases
RGD1309160
Chromosome Location
Chr. 1: 80417514 - 80424429 on Build Rnor_6.0
UniGene ID
Rn.19687
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109410.1 NP_001102880.1 3 233

View Full Product Details