Gene Transcripts

Gene Symbol
Dennd5a
Entrez Gene ID
308942
Gene Name
DENN/MADD domain containing 5A
Gene Aliases
Rab6ip1
Chromosome Location
Chr. 1: 174524047 - 174588529 on Build Rnor_6.0
UniGene ID
Rn.107101
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107546.2 NP_001101016.2 16 3047
GenBank mRNA BC168925.1 - 3 269

View Full Product Details