Gene Transcripts

Gene Symbol
Hmgb1
Entrez Gene ID
25459
Gene Name
high mobility group box 1
Gene Aliases
Ac2-008, Hmg1
Chromosome Location
Chr. 12: 7090927 - 7092190 on Build Rnor_6.0
UniGene ID
Rn.144565
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012963.2 NP_037095.1 2204
GenBank mRNA BC081839.1 - 2204

View Full Product Details