Gene Transcripts

Gene Symbol
Inafm1
Entrez Gene ID
100192314
Gene Name
InaF-motif containing 1
Gene Aliases
Prr24
Chromosome Location
Chr. 1: 78210866 - 78212350 on Build Rnor_6.0
UniGene ID
Rn.217950
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136272.1 NP_001129744.1 1 1092
GenBank mRNA AW144299.1 - 1 450

View Full Product Details