Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015319.2 | NP_056134.2 | 26 | 4191 | |
NM_170754.2 | NP_736610.2 | 26 | 4101 | ||
NM_198316.1 | NP_938072.1 | 26 | 3929 | ||
XM_006719302.3 | XP_006719365.1 | 26 | 5577 | ||
XM_006719303.1 | XP_006719366.1 | 26 | 4147 | ||
XM_011538079.1 | XP_011536381.1 | 26 | 3975 | ||
XM_017019088.1 | XP_016874577.1 | 26 | 4019 | ||
XM_017019089.1 | XP_016874578.1 | 26 | 4002 | ||
GenBank mRNA | AB028998.2 | - | 26 | 4256 | |
AF417490.1 | - | 26 | 3929 | ||
AF518728.1 | - | 26 | 4191 | ||
AF518729.1 | - | 26 | 4101 | ||
AK131320.1 | - | 19 | 2203 | ||
AL137564.1 | - | 9 | 1687 | ||
BC054099.1 | - | 12 | 2588 | ||
BC110854.1 | - | 20 | 3614 | ||
BC129828.1 | - | 21 | 3649 | 40114077 | |
BC129829.1 | - | 21 | 3649 | 40114082 | |
BC131503.1 | - | 3 | 290 | ||
BC142668.1 | - | 26 | 4084 | 40148166 | |
BC142712.1 | - | 26 | 4084 | 40148847 | |
BC150259.1 | - | 26 | 4256 | 9007193 | |
BX647126.1 | - | 27 | 4055 | ||
CR936725.1 | - | 26 | 4184 |