Gene Transcripts

Gene Symbol
Zfp637
Entrez Gene ID
362425
Gene Name
zinc finger protein 637
Gene Aliases
AZF1, DZF11
Chromosome Location
Chr. 4: 149857120 - 149882828 on Build Rnor_6.0
UniGene ID
Rn.63909
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134908.1 NP_001128380.1 3 218
XM_006237192.2 XP_006237254.1 4 905
XM_006237193.2 XP_006237255.1 4 934
XM_006237194.2 XP_006237256.1 4 860
GenBank mRNA BC158822.1 - 3 218
FQ213084.1 - 3 190

View Full Product Details