Gene Transcripts

Gene Symbol
Cox7c
Entrez Gene ID
100188937
Gene Name
cytochrome c oxidase, subunit VIIc
Gene Aliases
Cox7c1
Chromosome Location
Chr. 2: 14699878 - 14701903 on Build Rnor_6.0
UniGene ID
Rn.105968
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134705.1 NP_001128177.1 3 416
GenBank mRNA FQ217721.1 - 3 365
FQ217795.1 - 3 365
FQ221226.1 - 3 426
FQ223619.1 - 3 367

View Full Product Details