Gene Transcripts
- Gene Symbol
- PIP5K1C
- Entrez Gene ID
- 23396
- Gene Name
- phosphatidylinositol-4-phosphate 5-kinase type 1 gamma
- Gene Aliases
- LCCS3, PIP5K-GAMMA, PIP5K1-gamma, PIP5Kgamma
- Chromosome Location
- Chr. 19: 3630181 - 3700492 on Build GRCh38
- UniGene ID
- Hs.282177
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001195733.1 |
NP_001182662.1
|
9, 10 |
1298 |
|
NM_001300849.1 |
NP_001287778.1
|
9, 10 |
1311 |
|
NM_012398.2 |
NP_036530.1
|
9, 10 |
1298 |
|
XM_011527845.1 |
XP_011526147.1
|
9, 10 |
1305 |
|
XM_011527846.1 |
XP_011526148.1
|
9, 10 |
1305 |
|
XM_011527848.1 |
XP_011526150.1
|
9, 10 |
1305 |
|
XM_011527849.1 |
XP_011526151.1
|
9, 10 |
1305 |
|
XM_011527850.2 |
XP_011526152.1
|
9, 10 |
1264 |
|
XM_011527851.2 |
XP_011526153.1
|
2, 3 |
892 |
|
XM_011527853.2 |
XP_011526155.1
|
2, 3 |
723 |
|
XM_017026540.1 |
XP_016882029.1
|
9, 10 |
1305 |
|
GenBank mRNA |
AB011161.1 |
- |
9, 10 |
1266 |
|
AK301309.1 |
- |
9, 10 |
1281 |
|
AK315912.1 |
- |
9, 10 |
1284 |
|
FJ965536.1 |
- |
9, 10 |
1209 |
|
FJ965537.1 |
- |
9, 10 |
1209 |
|
View Full Product Details