Gene Transcripts

Gene Symbol
NCAPH
Entrez Gene ID
23397
Gene Name
non-SMC condensin I complex subunit H
Gene Aliases
BRRN1, CAP-H
Chromosome Location
Chr. 2: 96335741 - 96377095 on Build GRCh38
UniGene ID
Hs.308045
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001281710.1 NP_001268639.1 14 1886
NM_001281711.1 NP_001268640.1 14 1847
NM_001281712.1 NP_001268641.1 13 1666
NM_015341.4 NP_056156.2 14 1919
XM_005263908.3 XP_005263965.1 14 1859
XM_006712388.3 XP_006712451.1 14 1766
GenBank mRNA AK291053.1 - 14 1860
AK299248.1 - 14 1788
AK303725.1 - 13 1611
AK308050.1 - 14 1827
BC024211.2 - 14 1873 3504782
D38553.1 - 14 1849

View Full Product Details