Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001143775.1 | NP_001137247.1 | 3 | 586 | |
NM_015343.4 | NP_056158.2 | 4 | 653 | ||
GenBank mRNA | AJ011916.1 | - | 3 | 236 | |
AY364239.1 | - | 3 | 206 | ||
BC009295.2 | - | 4 | 652 | 4123279 |