Gene Transcripts
- Gene Symbol
- TRAM1
- Entrez Gene ID
- 23471
- Gene Name
- translocation associated membrane protein 1
- Gene Aliases
- PNAS8, TRAM, TRAMP
- Chromosome Location
- Chr. 8: 70573218 - 70608459 on Build GRCh38
- UniGene ID
- Hs.491988
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001317804.1 |
NP_001304733.1
|
4, 5 |
480 |
|
NM_001317805.1 |
NP_001304734.1
|
3, 4 |
995 |
|
NM_014294.5 |
NP_055109.1
|
3, 4 |
563 |
|
GenBank mRNA |
AK289397.1 |
- |
3, 4 |
423 |
|
AK292777.1 |
- |
3, 4 |
347 |
|
AK303411.1 |
- |
4, 5 |
408 |
|
BC000687.2 |
- |
3, 4 |
385 |
3347823 |
BC020283.1 |
- |
3, 4 |
378 |
|
BC037738.1 |
- |
3, 4 |
475 |
5261490 |
BT007359.1 |
- |
3, 4 |
303 |
|
CR541654.1 |
- |
3, 4 |
303 |
|
CR541739.1 |
- |
3, 4 |
303 |
|
X63679.1 |
- |
3, 4 |
424 |
|
View Full Product Details