Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012269.2 | NP_036401.2 | 3 | 736 | |
XM_011515990.2 | XP_011514292.1 | 3 | 812 | ||
XM_011515991.2 | XP_011514293.1 | 3 | 394 | ||
XM_017011911.1 | XP_016867400.1 | 4 | 439 | ||
GenBank mRNA | AB470346.1 | - | 2 | 168 | |
AF009010.1 | - | 3 | 739 | ||
BC104788.1 | - | 2 | 174 | 8143791 | |
BC104790.1 | - | 2 | 174 | 8143793 | |
BG013757.1 | - | 3 | 279 |