Gene Transcripts
- Gene Symbol
- CORO1C
- Entrez Gene ID
- 23603
- Gene Name
- coronin 1C
- Gene Aliases
- HCRNN4
- Chromosome Location
- Chr. 12: 108645109 - 108731550 on Build GRCh38
- UniGene ID
- Hs.330384
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001105237.2 |
NP_001098707.1
|
2, 3 |
405 |
|
NM_001276471.1 |
NP_001263400.1
|
2, 3 |
423 |
|
NM_014325.3 |
NP_055140.1
|
2, 3 |
324 |
|
XM_017019121.1 |
XP_016874610.1
|
3, 4 |
1153 |
|
XM_017019122.1 |
XP_016874611.1
|
2, 3 |
730 |
|
XM_017019123.1 |
XP_016874612.1
|
3, 4 |
783 |
|
GenBank mRNA |
AB030656.1 |
- |
2, 3 |
293 |
|
AB209909.1 |
- |
2, 3 |
279 |
|
AK023205.1 |
- |
2, 3 |
294 |
|
AK096363.1 |
- |
2, 3 |
423 |
|
AK223073.1 |
- |
2, 3 |
292 |
|
AK297463.1 |
- |
2, 3 |
268 |
|
AL162070.1 |
- |
2, 3 |
262 |
|
AM849477.1 |
- |
2, 3 |
298 |
|
AM849478.1 |
- |
2, 3 |
405 |
|
BC002342.2 |
- |
2, 3 |
278 |
2822459 |
DB450956.1 |
- |
2, 3 |
324 |
|
View Full Product Details