Gene Transcripts

Gene Symbol
ZIM2
Entrez Gene ID
23619
Gene Name
zinc finger imprinted 2
Gene Aliases
ZNF656
Chromosome Location
Chr. 19: 56774547 - 56840729 on Build GRCh38
UniGene ID
Hs.731875
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146326.1 NP_001139798.1 12 1329
NM_001146327.1 NP_001139799.1 12 1416
NM_015363.4 NP_056178.3 11 1253
GenBank mRNA AF166122.1 - 11 1236
BC104876.1 - 11 1160 8143879
BC104878.1 - 11 1244 8143881

View Full Product Details