Gene Transcripts

Gene Symbol
BAMBI
Entrez Gene ID
25805
Gene Name
BMP and activin membrane bound inhibitor
Gene Aliases
NMA
Chromosome Location
Chr. 10: 28677495 - 28682939 on Build GRCh38
UniGene ID
Hs.533336
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012342.2 NP_036474.1 2 569
GenBank mRNA AK223065.1 - 2 541
BC019252.1 - 2 554 3503549
BT006820.1 - 2 166
U23070.1 - 2 538

View Full Product Details