Gene Transcripts

Gene Symbol
NDUFAF3
Entrez Gene ID
25915
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 3
Gene Aliases
2P1, C3orf60, E3-3
Chromosome Location
Chr. 3: 49020475 - 49023495 on Build GRCh38
UniGene ID
Hs.31387
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199069.1 NP_951032.1 5 1250
NM_199070.1 NP_951033.1 5 813
NM_199073.1 NP_951047.1 5 884
NM_199074.1 NP_951056.1 5 765

View Full Product Details