Gene Transcripts

Gene Symbol
CCIN
Entrez Gene ID
881
Gene Name
calicin
Gene Aliases
BTBD20, KBTBD14
Chromosome Location
Chr. 9: 36169392 - 36171334 on Build GRCh38
UniGene ID
Hs.115460
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005893.2 NP_005884.2 1 474
GenBank mRNA AF333334.1 - 1 472
AK313763.1 - 1 479
BC019251.2 - 1 442 4825221
Z46967.1 - 1 354

View Full Product Details