Gene Transcripts
- Gene Symbol
- UBXN7
- Entrez Gene ID
- 26043
- Gene Name
- UBX domain protein 7
- Gene Aliases
- UBXD7
- Chromosome Location
- Chr. 3: 196347659 - 196454323 on Build GRCh38
- UniGene ID
- Hs.518524
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_015562.1 |
NP_056377.1
|
4, 5 |
428 |
|
XM_011512670.2 |
XP_011510972.1
|
4, 5 |
376 |
|
XM_011512671.2 |
XP_011510973.1
|
3, 4 |
233 |
|
GenBank mRNA |
AB018337.1 |
- |
4, 5 |
356 |
|
AK026451.1 |
- |
4, 5 |
381 |
|
AK129880.1 |
- |
4, 5 |
381 |
|
AK303630.1 |
- |
3, 4 |
233 |
|
BC028986.1 |
- |
3, 4 |
260 |
|
BC047420.1 |
- |
4, 5 |
378 |
|
DA119173.1 |
- |
4, 5 |
428 |
|
View Full Product Details