Gene Transcripts

Gene Symbol
TBC1D29
Entrez Gene ID
26083
Gene Name
TBC1 domain family member 29
Gene Aliases
Chromosome Location
Chr. 17: 30559566 - 30565667 on Build GRCh38
UniGene ID
Hs.241421
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015594.2 NP_056409.1 5 538
XM_011524596.1 XP_011522898.1 5 464
GenBank mRNA AL117394.1 - 6 2887
BC096718.1 - 5 538 40003088
BC098126.1 - 4 489 40003085

View Full Product Details