Gene Transcripts

Gene Symbol
PTPN20
Entrez Gene ID
26095
Gene Name
protein tyrosine phosphatase, non-receptor type 20
Gene Aliases
CT126, PTPN20A, PTPN20B, bA142I17.1, bA42B19.1
Chromosome Location
Chr. 10: 46911134 - 47003834 on Build GRCh38
UniGene ID
Hs.440733
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042357.4 NP_001035816.1 10, 11 1418
NM_001042358.4 NP_001035817.1 9, 10 1276
NM_001042359.3 NP_001035818.1 7, 8 850
NM_001042360.3 NP_001035819.1 4, 5 727
NM_001042361.4 NP_001035820.1 7, 8 725
NM_001042362.3 NP_001035821.1 5, 6 482
NM_001042363.4 NP_001035822.1 9, 10 1261
NM_001042364.4 NP_001035823.1 6, 7 615
NM_001042365.3 NP_001035824.1 3, 4 274
NM_001320681.1 NP_001307610.1 8, 9 1166
NM_001320682.1 NP_001307611.1 4, 5 372
NM_001320683.1 NP_001307612.1 8, 9 882
NM_001320684.1 NP_001307613.1 7, 8 791
NM_001320685.1 NP_001307614.1 9, 10 1338
NM_001320686.1 NP_001307615.1 8, 9 1181
NM_001320688.1 NP_001307617.1 6, 7 755
NM_001320689.1 NP_001307618.1 5, 6 523
NM_001320690.1 NP_001307619.1 4, 5 387
NM_001320691.1 NP_001307620.1 7, 8 1090
NM_015605.8 NP_056420.3 8, 9 867
XM_011539605.2 XP_011537907.1 10, 11 1562
XM_011539606.2 XP_011537908.1 10, 11 2654
XM_011539607.2 XP_011537909.1 10, 11 1553
XM_011539610.2 XP_011537912.1 7, 8 1036
XM_011539611.2 XP_011537913.1 6, 7 1036
XM_017016042.1 XP_016871531.1 10, 11 1397
XM_017016044.1 XP_016871533.1 9, 10 1319
XM_017016045.1 XP_016871534.1 4, 5 2638
XM_017016046.1 XP_016871535.1 9, 10 1181
XM_017016047.1 XP_016871536.1 9, 10 1181
XM_017016049.1 XP_016871538.1 7, 8 912
XM_017016050.1 XP_016871539.1 5, 6 779
XM_017016052.1 XP_016871541.1 8, 9 846
XM_017016053.1 XP_016871542.1 7, 8 755
XM_017016054.1 XP_016871543.1 7, 8 768
XM_017016055.1 XP_016871544.1 7, 8 755
XM_017016056.1 XP_016871545.1 6, 7 763
XM_017016057.1 XP_016871546.1 7, 8 1073
XM_017016058.1 XP_016871547.1 5, 6 799
XM_017016059.1 XP_016871548.1 6, 7 757
XM_017016060.1 XP_016871549.1 6, 7 603
XM_017016061.1 XP_016871550.1 5, 6 553
XM_017016062.1 XP_016871551.1 5, 6 673
XM_017016063.1 XP_016871552.1 6, 7 644
XM_017016064.1 XP_016871553.1 4, 5 396
XM_017016065.1 XP_016871554.1 5, 6 446
GenBank mRNA AL050040.1 - 8, 9 1124
AY704141.1 - 9, 10 1217
AY704142.1 - 7, 8 791
AY704143.1 - 4, 5 668
AY704144.1 - 7, 8 666
AY704145.1 - 5, 6 423
AY704146.1 - 9, 10 1266
AY704147.1 - 8, 9 823
AY704148.1 - 7, 8 732
AY704149.1 - 5, 6 472
AY704150.1 - 9, 10 1279
AY704151.1 - 6, 7 620
AY704152.1 - 8, 9 1122
AY704153.1 - 6, 7 696
AY704154.1 - 6, 7 571
AY704155.1 - 5, 6 513
AY704156.1 - 5, 6 464
AY704157.1 - 4, 5 328
AY704158.1 - 7, 8 1031
BC093750.1 - 7, 8 721 7939595
BC113473.1 - 7, 8 721 8322525
BC143394.1 - 5, 6 478
BC171725.1 - 5, 6 478
BX648913.1 - 3, 4 275
CR933668.1 - 4, 5 340

View Full Product Details