Gene Transcripts

Gene Symbol
KIF1BP
Entrez Gene ID
26128
Gene Name
KIF1 binding protein
Gene Aliases
KBP, KIAA1279, TTC20
Chromosome Location
Chr. 10: 68988721 - 69016983 on Build GRCh38
UniGene ID
Hs.279580
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015634.3 NP_056449.1 4 843
GenBank mRNA AB033105.1 - 4 766
AK291343.1 - 4 748
BC006427.1 - 4 766
BC012180.1 - 4 740 4550085

View Full Product Details